Learn More
Selleck Chemical LLC ()-Dizocilpine (MK 801) Maleate S2876-1g
SDP
Catalog No. 503112312
Encompass_Preferred
( )-Dizocilpine (MK 801) Maleate is a potent selective and non-competitive NMDA receptor antagonist with Kd of 37 2 nM in rat brain membranes ( )-Dizocilpine (MK 801) Maleate can be used to induce animal models of Schizophrenia
Catalog No. 50-311-2312 Supplier Selleck Chemical LLC Supplier No. S28761G
( )-Dizocilpine (MK 801) Maleate is a potent selective and non-competitive NMDA receptor antagonist with Kd of 37 2 nM in rat brain membranes ( )-Dizocilpine (MK 801) Maleate can be used to induce animal models of Schizophrenia