Learn More
Medchemexpress LLC Dizocilpine maleate | 77086-22-7 | 337.37 | 50 MG
SDP
Catalog No. 5000366203
Encompass_Preferred
Dizocilpine maleate (MK-801 maleate) is a potent, selective, and non-competitive NMDA receptor antagonist with a Kd of 37.2 nM in rat brain membranes. It causes a progressive, long-lasting blockade of current induced by N-Me-D-Asp and can inhibit the activation of microglia induced by LPS. It is also used to induce schizophrenia models.
  • Potent, selective, and non-competitive NMDA receptor antagonist
  • Binds with NMDA receptor
  • Blocks N-Me-D-Asp-activated single-channel activity
  • Inhibits activation of microglia induced by LPS
  • Can be used to induce schizophrenia models
Catalog No. 50-003-66203 Supplier Medchemexpress LLC Supplier No. 50MGHY15084
Dizocilpine maleate (MK-801 maleate) is a potent, selective, and non-competitive NMDA receptor antagonist with a Kd of 37.2 nM in rat brain membranes. It causes a progressive, long-lasting blockade of current induced by N-Me-D-Asp and can inhibit the activation of microglia induced by LPS. It is also used to induce schizophrenia models.
  • Potent, selective, and non-competitive NMDA receptor antagonist
  • Binds with NMDA receptor
  • Blocks N-Me-D-Asp-activated single-channel activity
  • Inhibits activation of microglia induced by LPS
  • Can be used to induce schizophrenia models