Learn More
Medchemexpress LLC Dizocilpine maleate | 77086-22-7 | C20H19NO4 | 1 ML
SDP
Catalog No. 5000375862
Encompass_Preferred
Dizocilpine maleate (MK-801 maleate) is a potent, selective, and non-competitive antagonist of the NMDA receptor. It has a Kd of 37.2 nM in rat brain membranes and causes a progressive, long-lasting blockade of N-Me-D-Asp and NMDA-induced currents.
  • Potent, selective, and non-competitive NMDA receptor antagonist
  • Blocks NMDA-activated single-channel activity
  • Inhibits activation of microglia induced by LPS
  • Reduces microglial TNF-α output
  • Can be used to induce schizophrenia models
Catalog No. 50-003-75862 Supplier Medchemexpress LLC Supplier No. HY1508410MM/1ML
Dizocilpine maleate (MK-801 maleate) is a potent, selective, and non-competitive antagonist of the NMDA receptor. It has a Kd of 37.2 nM in rat brain membranes and causes a progressive, long-lasting blockade of N-Me-D-Asp and NMDA-induced currents.
  • Potent, selective, and non-competitive NMDA receptor antagonist
  • Blocks NMDA-activated single-channel activity
  • Inhibits activation of microglia induced by LPS
  • Reduces microglial TNF-α output
  • Can be used to induce schizophrenia models