Learn More
Medchemexpress LLC Dizocilpine maleate | 77086-22-7 | C20H19NO4 | 100 MG
SDP
Catalog No. 5000375468
Encompass_Preferred
Dizocilpine maleate is a potent, selective, and non-competitive NMDA receptor antagonist. It effectively blocks currents induced by N-Me-D-Asp and can be utilized in various research applications, including inducing schizophrenia models.
  • Potent, selective, and non-competitive NMDA receptor antagonist
  • Causes progressive, long-lasting blockade of current induced by N-Me-D-Asp
  • Used to induce schizophrenia models
  • Kd of 37.2 nM in rat brain membranes
Catalog No. 50-003-75468 Supplier Medchemexpress LLC Supplier No. HY15084100MG
Dizocilpine maleate is a potent, selective, and non-competitive NMDA receptor antagonist. It effectively blocks currents induced by N-Me-D-Asp and can be utilized in various research applications, including inducing schizophrenia models.
  • Potent, selective, and non-competitive NMDA receptor antagonist
  • Causes progressive, long-lasting blockade of current induced by N-Me-D-Asp
  • Used to induce schizophrenia models
  • Kd of 37.2 nM in rat brain membranes