Learn More
Medchemexpress LLC Dizocilpine maleate | 77086-22-7 | C20H19NO4 | 25 MG
SDP
Catalog No. 5000374500
Encompass_Preferred
Dizocilpine maleate (MK-801 maleate) is a potent, selective, and non-competitive NMDA receptor antagonist. It effectively blocks N-Me-D-Asp-induced current and inhibits microglial activation, reducing TNF-α output. This compound is also utilized for inducing schizophrenia models in rats.
  • Potent, selective, and non-competitive NMDA receptor antagonist
  • Blocks current induced by N-Me-D-Asp
  • Inhibits activation of microglia induced by LPS
  • Reduces microglial TNF-α output
  • Used to induce schizophrenia models in rats
  • Cited in over 40 publications
Catalog No. 50-003-74500 Supplier Medchemexpress LLC Supplier No. 25MGHY15084
Dizocilpine maleate (MK-801 maleate) is a potent, selective, and non-competitive NMDA receptor antagonist. It effectively blocks N-Me-D-Asp-induced current and inhibits microglial activation, reducing TNF-α output. This compound is also utilized for inducing schizophrenia models in rats.
  • Potent, selective, and non-competitive NMDA receptor antagonist
  • Blocks current induced by N-Me-D-Asp
  • Inhibits activation of microglia induced by LPS
  • Reduces microglial TNF-α output
  • Used to induce schizophrenia models in rats
  • Cited in over 40 publications