Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dizocilpine (maleate) | 77086-22-7 | C20H19NO4 | 5 MG
SDP

Catalog No. 5000374952
Encompass_Preferred

Dizocilpine maleate (MK-801 maleate) is a potent, selective, and non-competitive NMDA receptor antagonist. It targets iGluR, involved in Membrane Transporter/Ion Channel and Neuronal Signaling pathways. This compound has a Kd of 37.2 nM in rat brain membranes and a molecular weight of 337.37.

  • Potent, selective, non-competitive NMDA receptor antagonist
  • Targets iGluR, Membrane Transporter/Ion Channel, and Neuronal Signaling
  • Soluble in DMSO, ethanol, and water (requires ultrasonic)
  • Can be used to induce schizophrenia models
  • Inhibits activation of microglia induced by LPS
  • Reduces microglial TNF-α output
  • Recommended storage at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 2 years or -20°C for 1 year

Catalog No. 50-003-74952 Supplier Medchemexpress LLC Supplier No. HY150845MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dizocilpine maleate (MK-801 maleate) is a potent, selective, and non-competitive NMDA receptor antagonist. It targets iGluR, involved in Membrane Transporter/Ion Channel and Neuronal Signaling pathways. This compound has a Kd of 37.2 nM in rat brain membranes and a molecular weight of 337.37.

  • Potent, selective, non-competitive NMDA receptor antagonist
  • Targets iGluR, Membrane Transporter/Ion Channel, and Neuronal Signaling
  • Soluble in DMSO, ethanol, and water (requires ultrasonic)
  • Can be used to induce schizophrenia models
  • Inhibits activation of microglia induced by LPS
  • Reduces microglial TNF-α output
  • Recommended storage at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 2 years or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.