Learn More
Enzo Life Sciences DJ 1 monoclonal antibody (3E8) (100 µg)
SDP
Supplier:  Enzo Life Sciences ADIKAMSA100E
Encompass_Preferred
Clone: 3E8. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human DJ-1. UniProt: Q99497. Species Reactivity: Human. Applications: WB. Recommended dilutions: Western Blot (1-10 µg/ml, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-666-480