Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DJ-V-159 | 2253744-53-3 | 99.9% | 502.37 | 1 ML
SDP

Catalog No. 5000435974
Encompass_Preferred

DJ-V-159 is an agonist for G protein-coupled receptor family C group 6 member A (GPRC6A). It activates ERK and stimulates cAMP production in GPRC6A expressing HEK-293 cells. Additionally, it stimulates insulin secretion in mouse beta-cell MIN-6 cells and reduces blood glucose levels in wild-type mice.

  • Activates ERK in HEK-293 cells transfected with GPRC6A.
  • Stimulates cAMP production in GPRC6A expressing HEK-293 cells.
  • Stimulates insulin secretion in mouse beta-cell MIN-6 cells.
  • Reduces blood glucose levels in wild-type mice at a dose of 10 mg/kg.
  • Appearance: solid, white to off-white.
  • Soluble in DMSO (25 mg/mL).

Catalog No. 50-004-35974 Supplier Medchemexpress LLC Supplier No. HY11416510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DJ-V-159 is an agonist for G protein-coupled receptor family C group 6 member A (GPRC6A). It activates ERK and stimulates cAMP production in GPRC6A expressing HEK-293 cells. Additionally, it stimulates insulin secretion in mouse beta-cell MIN-6 cells and reduces blood glucose levels in wild-type mice.

  • Activates ERK in HEK-293 cells transfected with GPRC6A.
  • Stimulates cAMP production in GPRC6A expressing HEK-293 cells.
  • Stimulates insulin secretion in mouse beta-cell MIN-6 cells.
  • Reduces blood glucose levels in wild-type mice at a dose of 10 mg/kg.
  • Appearance: solid, white to off-white.
  • Soluble in DMSO (25 mg/mL).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.