Learn More
Medchemexpress LLC Dj-v-159 | 2253744-53-3 | 99.9% | 502.37 | 100 MG
SDP
Catalog No. 5000436846
Encompass_Preferred
DJ-V-159 is an agonist for G protein-coupled receptor family C group 6 member A (GPRC6A).
  • Activates ERK in HEK-293 cells transfected with GPRC6A.
  • Dose-dependently stimulates cAMP production in GPRC6A expressing HEK-293 cells.
  • Stimulates insulin secretion in mouse beta-cell MIN-6 cells.
  • Reduces blood glucose levels in wildtype mice at a dose of 10 mg/kg.
Catalog No. 50-004-36846 Supplier Medchemexpress LLC Supplier No. HY114165100MG
DJ-V-159 is an agonist for G protein-coupled receptor family C group 6 member A (GPRC6A).
  • Activates ERK in HEK-293 cells transfected with GPRC6A.
  • Dose-dependently stimulates cAMP production in GPRC6A expressing HEK-293 cells.
  • Stimulates insulin secretion in mouse beta-cell MIN-6 cells.
  • Reduces blood glucose levels in wildtype mice at a dose of 10 mg/kg.