Learn More
Medchemexpress LLC DJ-V-159 | 2253744-53-3 | 99.9% | 502.37 | 50 MG
SDP
Catalog No. 5000437319
Encompass_Preferred
DJ-V-159 is an agonist for G protein-coupled receptor family C group 6 member A (GPRC6A). It activates ERK in HEK-293 cells transfected with GPRC6A and dose-dependently stimulates cAMP production. DJ-V-159 also stimulates insulin secretion in mouse beta-cell MIN-6 cells, similar to the effects of Ocn, a known ligand of GPRC6A, and has been shown to reduce blood glucose levels in wildtype mice.
  • Activates ERK in HEK-293 cells transfected with GPRC6A.
  • Stimulates cAMP production in GPRC6A expressing HEK-293 cells.
  • Stimulates insulin secretion in mouse beta-cell MIN-6 cells.
  • Reduces blood glucose levels in wildtype mice.
  • For research use only.
Catalog No. 50-004-37319 Supplier Medchemexpress LLC Supplier No. HY11416550MG
DJ-V-159 is an agonist for G protein-coupled receptor family C group 6 member A (GPRC6A). It activates ERK in HEK-293 cells transfected with GPRC6A and dose-dependently stimulates cAMP production. DJ-V-159 also stimulates insulin secretion in mouse beta-cell MIN-6 cells, similar to the effects of Ocn, a known ligand of GPRC6A, and has been shown to reduce blood glucose levels in wildtype mice.
  • Activates ERK in HEK-293 cells transfected with GPRC6A.
  • Stimulates cAMP production in GPRC6A expressing HEK-293 cells.
  • Stimulates insulin secretion in mouse beta-cell MIN-6 cells.
  • Reduces blood glucose levels in wildtype mice.
  • For research use only.