Learn More
Medchemexpress LLC Djalonensone-d3 | 2468774-90-3 | 275.27 | C15H9D3O5 | 1 MG
SDP
Catalog No. 5000303232
Encompass_Preferred
Djalonensone-d3 is a deuterium-labeled analog of djalonensone (CAS 2468774-90-3; molecular formula C15H9D3O5; molecular weight 275.27). Supplied in small analytical quantities, it is intended for use as a tracer and as an internal standard in quantitative NMR, GC-MS, and LC-MS workflows.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Provided in small, precisely weighed quantities suitable for reference use.
  • Useful as a tracer in metabolic and analytical studies.
Catalog No. 50-003-03232 Supplier Medchemexpress LLC Supplier No. HYW013863S1MG
Djalonensone-d3 is a deuterium-labeled analog of djalonensone (CAS 2468774-90-3; molecular formula C15H9D3O5; molecular weight 275.27). Supplied in small analytical quantities, it is intended for use as a tracer and as an internal standard in quantitative NMR, GC-MS, and LC-MS workflows.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Provided in small, precisely weighed quantities suitable for reference use.
  • Useful as a tracer in metabolic and analytical studies.