Learn More
Medchemexpress LLC HY-P7154 10ug Medchemexpress, DKK-1 (CHO-expressed), Mouse CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP715410UG
Encompass_Preferred
Medchemexpress, HY-P7154 10ug DKK-1 (CHO-expressed), Mouse CAS: Recombinant Mouse DKK-1 (CHO-expressed) is shown to be a potent inhibitor of Wnt signaling and member of dickkopf family. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4353