Learn More
Enzo Life Sciences Dkk-1 (mouse), (recombinant) (active) (5µg)
SDP
Supplier:  Enzo Life Sciences ENZ60002C005
Encompass_Preferred
Alternative name: Dickkopf-related protein-1. Purity: ≥80% (SDS-PAGE). Formulation: Lyophilized from a solution containing 1X PBS, 1% CHAPS, and 0.1% BSA. Source: Produced in insect cells. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-2655