Learn More
ACROBiosystems ACROBiosystems Mouse Dkk-3 Protein, His Tag
Supplier:  ACROBiosystems DK3M52261MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse Dkk-3, His Tag (DK3-M5226) is expressed from human 293 cells (HEK293). It contains AA Pro 23 - Ile 349 (Accession # NP_056629.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.1 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7249