Learn More
Genscript Corporation DKK1 C terminal Domain hFc Chi
Catalog No. 5000059093
Encompass
Dickkopf-1 (Dkk1) the founding and best-studied member of the Dkk family functions as an antagonist of canonical Wnt/ -catenin Dkk1 is considered to play a broad role in a variety of biological processes
Catalog No. 50-000-59093 Supplier Genscript Corporation Supplier No. Z04438100
Dickkopf-1 (Dkk1) the founding and best-studied member of the Dkk family functions as an antagonist of canonical Wnt/ -catenin Dkk1 is considered to play a broad role in a variety of biological processes