Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ DKK1 (Human) ELISA Kit

Catalog No. 89101035 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Plasma (EDTA, Heparin), Serum
CCS, Plasma, Serum, Urine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-101-035 CCS, Plasma (EDTA, Heparin), Serum
89-028-580 CCS, Plasma, Serum, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-101-035 Supplier Abnova™ Supplier No. KA3356
Add to Cart
Add to Cart

In-vitro ELISA for the quantitative measurement of Human DKK1

Specifications

Calibration Range 62.5 to 4000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description DKK1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human DKK1.
Detection Method Colorimetric
Includes Component, 96-well plate precoated with anti- human DKK1 antibody, Lyophilized recombinant human DKK1 standard (10ng x 2), Biotinylated anti- human DKK1 antibody(dilution 1:100) (130μL), Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100) (130μL), Sample diluent buffer (30mL), Antibody diluent buffer (12mL), ABC diluent buffer (12mL), TMB color developing agent (10mL), TMB stop solution (10mL)
Quantity 1 Kit
Sample Type CCS, Plasma (EDTA, Heparin), Serum
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type DKK1 (Human) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.