Human Dkk-1 protein is a member of the dickkopf-related protein family that plays a vital role in embryonic development by inhibiting the WNT signaling pathway. By binding to LRP6 with high affinity, this protein prevents Frizzled-Wnt-LRP6 complex formation and promotes LRP6 internalization and degradation via Kremen. It is also involved in joint remodeling and bone formation, with implications in the pathogenesis of rheumatoid arthritis and multiple myeloma. This protein is expressed in HEK293 cells and contains a human IgG1 Fc tag at the C-terminus.
- Greater than 95% purity by SDS-PAGE and greater than 90% purity by SEC-MALS
- Calculated molecular weight of 52.4 kDa, migrating at 67-85 kDa under reducing conditions due to glycosylation
- Lyophilized from a 0.22 μm filtered PBS solution, pH 7.4, containing trehalose
- Endotoxin level less than 1.0 EU per μg