Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human Dkk-1 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;DK1,DKK1,DKK-1,SK
Catalog No. 501657857
Encompass_Preferred
Recombinant Protein;Biotinylated Human Dkk-1 Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human Dkk-1, Fc Tag, Avi Tag (DK1-H82F5) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 266 (Accession # O94907-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 54.0 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7857 Supplier ACROBiosystems Supplier No. DK1H82F525UG
Recombinant Protein;Biotinylated Human Dkk-1 Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human Dkk-1, Fc Tag, Avi Tag (DK1-H82F5) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 266 (Accession # O94907-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 54.0 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.