Learn More
ACROBiosystems Recombinant Protein;Human Dkk-1 Protein;HEK293;100UG;DK1,DKK1,DKK-1,SK
Catalog No. 501657538
Encompass_Preferred
Recombinant Protein;Human Dkk-1 Protein;100UG;Human Dkk-1, His Tag (DK1-H5221) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 266 (Accession # NP_036374.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.8 kDa. The protein migrates as 40-46 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7538 Supplier ACROBiosystems Supplier No. DK1H5221100UG
Recombinant Protein;Human Dkk-1 Protein;100UG;Human Dkk-1, His Tag (DK1-H5221) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 266 (Accession # NP_036374.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.8 kDa. The protein migrates as 40-46 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.