Learn More
ACROBiosystems Recombinant Protein;Mouse Dkk-1 Protein;HEK293;500UG;DK1,DKK1,DKK-1,SK
Catalog No. 501658732
Encompass_Preferred
Recombinant Protein;Mouse Dkk-1 Protein;500UG;Mouse Dkk-1, His Tag (DK1-M52H6) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 272 (Accession # O54908).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28 kDa. The protein migrates as 41-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8732 Supplier ACROBiosystems Supplier No. DK1M52H6500UG
Recombinant Protein;Mouse Dkk-1 Protein;500UG;Mouse Dkk-1, His Tag (DK1-M52H6) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 272 (Accession # O54908).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28 kDa. The protein migrates as 41-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.