Learn More
ACROBiosystems Recombinant Protein;Human Dkk-3 Protein;HEK293;200UG;DK3,DKK3,DKK-3,REIC,RIG
Supplier:  ACROBiosystems DK3H5220200UG
Encompass_Preferred
Recombinant Protein;Human Dkk-3 Protein;200UG;Human Dkk-3, His Tag (DK3-H5220) is expressed from human 293 cells (HEK293). It contains AA Pro 23 - Ile 350 (Accession # NP_001018067.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.6 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7541