Learn More
ACROBiosystems Recombinant Protein;Mouse Dkk-3 Protein;HEK293;50UG;DK3,DKK3,DKK-3,REIC,RIG
Catalog No. 501658730
Encompass_Preferred
Recombinant Protein;Mouse Dkk-3 Protein;50UG;Mouse Dkk-3, His Tag (DK3-M5226) is expressed from human 293 cells (HEK293). It contains AA Pro 23 - Ile 349 (Accession # NP_056629.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.1 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8730 Supplier ACROBiosystems Supplier No. DK3M522650UG
Recombinant Protein;Mouse Dkk-3 Protein;50UG;Mouse Dkk-3, His Tag (DK3-M5226) is expressed from human 293 cells (HEK293). It contains AA Pro 23 - Ile 349 (Accession # NP_056629.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.1 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.