Learn More
Aviva Systems Biology Corp Dkk4 Antibody - C-terminal region (ARP42406_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42406_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dkk4. It was validated on Western Blot. . Alternate names: Dkk-4, MGC25705ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-000