Learn More
Medchemexpress LLC Dkm 2-93 | 65836-72-8 | MFCD01733639 | 98.8% | 243.69 | C11H14ClNO3 | 250 MG
SDP
Catalog No. 5000249302
Encompass_Preferred
DKM 2-93 is a covalent, relatively selective inhibitor of ubiquitin-like modifier activating enzyme 5 (UBA5) used for biochemical and cellular research. It inhibits UBA5 by covalently modifying the catalytic cysteine (reported biochemical IC50 ≈ 430 μM) and reduces viability of PaCa-2 and Panc-1 cells in cellular assays. The compound is supplied as a solid or as DMSO solutions; reported purity is 98.76% and molecular weight is 243.69.
Catalog No. 50-002-49302 Supplier Medchemexpress LLC Supplier No. HY101836250MG
DKM 2-93 is a covalent, relatively selective inhibitor of ubiquitin-like modifier activating enzyme 5 (UBA5) used for biochemical and cellular research. It inhibits UBA5 by covalently modifying the catalytic cysteine (reported biochemical IC50 ≈ 430 μM) and reduces viability of PaCa-2 and Panc-1 cells in cellular assays. The compound is supplied as a solid or as DMSO solutions; reported purity is 98.76% and molecular weight is 243.69.