Learn More
Bioworld DKW Basal Medium, w/ 30 g/L Sucrose, 50 L
Catalog No. 50254898
Encompass_Preferred
DKW Basal MediumContains Macronutrients, Micronutrients, and Sucrose as described by Driver & Kuniyuki (1984) and McGranaha et al. (1987)Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 35.22 g/LpH: 3.5–4.5Soluble in water
Catalog No. 50-254-898 Supplier Bioworld Supplier No. 306301614
DKW Basal MediumContains Macronutrients, Micronutrients, and Sucrose as described by Driver & Kuniyuki (1984) and McGranaha et al. (1987)Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 35.22 g/LpH: 3.5–4.5Soluble in water