Learn More
Bioworld DKW Basal Salt Mixture, 25 L
Catalog No. 50254748
Encompass_Preferred
DKW/Juglans Basal Salt MixtureContains Macronutrients and Micronutrients as described by Driver & Kuniyuki (1984) and McGranahan et al. (1987)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 5.22 g/LpH: 3.5–4.5Soluble in water
Catalog No. 50-254-748 Supplier Bioworld Supplier No. 306300923
DKW/Juglans Basal Salt MixtureContains Macronutrients and Micronutrients as described by Driver & Kuniyuki (1984) and McGranahan et al. (1987)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 5.22 g/LpH: 3.5–4.5Soluble in water