Learn More
PHYTOTECHNOLOGY LABS DKW BASAL SALT MIXTURE 100L
Catalog No. NC1789698
Encompass_Preferred
This medium was developed for the multiplication of shoots from nodal explants. The medium was supplemented with 4.5 μM BA and 5 nM IBA. Rooting the shoots was enhanced by dipping the basal ends of the shoots in 5 mM IBA prior to transferring to the greenhouse.
Catalog No. NC1789698 Supplier PHYTOTECHNOLOGY LABS Supplier No. D190100L
This medium was developed for the multiplication of shoots from nodal explants. The medium was supplemented with 4.5 μM BA and 5 nM IBA. Rooting the shoots was enhanced by dipping the basal ends of the shoots in 5 mM IBA prior to transferring to the greenhouse.