Learn More
PLANT CELL TECHNOLOGY DKW Basal Salts w/ Macro- and Micronutrients, Plant Tissue Culture, DKW Basal Salts Mod. 1, Described by Driver and Kuniyuki (1984) and McGranahan et al. (1987), Off-White to Yellow Powder, 50L Bottle
Catalog No. NC3905201
Encompass_Preferred
DKW Basal Salts Mod. 1 is formulated to provide essential nutrients for plant tissue culture of woody plant species. Supplied in a screw-top bottle, this 50L equivalent of off-white to yellow powder is ideal for use in research labs, commercial propagation, and educational applications. Stable under recommended storage conditions, ensuring reproducible tissue culture results.
Catalog No. NC3905201 Supplier PLANT CELL TECHNOLOGY Supplier No. DKP0150LT
DKW Basal Salts Mod. 1 is formulated to provide essential nutrients for plant tissue culture of woody plant species. Supplied in a screw-top bottle, this 50L equivalent of off-white to yellow powder is ideal for use in research labs, commercial propagation, and educational applications. Stable under recommended storage conditions, ensuring reproducible tissue culture results.