Learn More
PLANT CELL TECHNOLOGY DKW Basal Salts w/ Macro- and Micronutrients, Plant Tissue Culture, DKW Basal Salts Mod. 2, Described by Driver and Kuniyuki (1984) and McGranahan et al. (1987), Off-White to Yellow Powder, 10L Bottle
Catalog No. NC3905205
Encompass_Preferred
DKW Basal Salts Mod. 2 is formulated to provide essential nutrients and vitamins for plant tissue culture of woody plant species. Supplied in a screw-top bottle, this 10L equivalent of off white to yellow powder is ideal for use in research labs, commercial propagation, and educational applications. Stable under recommended storage conditions, ensuring reproducible tissue culture results.
Catalog No. NC3905205 Supplier PLANT CELL TECHNOLOGY Supplier No. DKP0210LT
DKW Basal Salts Mod. 2 is formulated to provide essential nutrients and vitamins for plant tissue culture of woody plant species. Supplied in a screw-top bottle, this 10L equivalent of off white to yellow powder is ideal for use in research labs, commercial propagation, and educational applications. Stable under recommended storage conditions, ensuring reproducible tissue culture results.