Learn More
Bioworld DKW/Juglans Medium w/ Vitamins, 25 L
Catalog No. 50254589
Encompass_Preferred
DKW/Juglans MediumContains Macronutrients, Micronutrients, and Vitamins as described by Driver & Kuniyuki (1984) and McGranahan et al. (1987) Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-589 Supplier Bioworld Supplier No. 306300203
DKW/Juglans MediumContains Macronutrients, Micronutrients, and Vitamins as described by Driver & Kuniyuki (1984) and McGranahan et al. (1987) Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested