Learn More
Bioworld DKW/Juglans Vitamin Mix (Concentrate), 100 mL
Supplier:  Bioworld 306300221
Encompass_Preferred
DKW/Juglans Vitamin MixContains Vitamins as described by Driver & Kuniyuki (1984) and McGranahan et al. (1987)Package contains 10.50 g vitamins to prepare 100 mL of 1000X vitamin stock solution Use 1 mL per liter of medium Contents: Glycine 2.00 mg/Lmyo-Inositol 100 mg/LNicotinic Acid 1 mg/LThiamine HCl 2 mg/L
Catalog No. 50-254-592