Learn More
Jackson Immuno Research Labs Alexa Fluor® 647 AffiniPure Donkey Anti-Rabbit IgG (H+L) 0.5mg
SDP
Catalog No. NC0254454
Encompass_Preferred
Whole IgG antibodies are isolated as intact molecules from antisera by immunoaffinity chromatography. They have an Fc portion and two antigen binding Fab portions joined together by disulfide bonds and therefore they are divalent. The average molecular weight is reported to be about 160 kDa. The whole IgG form of antibodies is suitable for the majority of immunodetection procedures and is the most cost effective. Purity: The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. Buffer: 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 Stabilizer: 15 mg/ml Bovine Serum Albumin (IgG-Free, Protease-Free) Preservative: 0.05% Sodium Azide
Catalog No. NC0254454 Supplier Jackson Immuno Research Labs Supplier No. 711605152
Whole IgG antibodies are isolated as intact molecules from antisera by immunoaffinity chromatography. They have an Fc portion and two antigen binding Fab portions joined together by disulfide bonds and therefore they are divalent. The average molecular weight is reported to be about 160 kDa. The whole IgG form of antibodies is suitable for the majority of immunodetection procedures and is the most cost effective. Purity: The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. Buffer: 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 Stabilizer: 15 mg/ml Bovine Serum Albumin (IgG-Free, Protease-Free) Preservative: 0.05% Sodium Azide