Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dl-01 formic | 2964513-44-6 | 98.5% | 1144.30 | C56H73N9O15S | 5 MG
SDP

Catalog No. 5000204411
Encompass_Preferred

DL-01 formic is a drug-linker conjugate used in the synthesis of antibody-drug conjugates (ADCs) and related research applications. It is supplied as a high-purity solid for laboratory research use only and should be handled and stored according to the product datasheet and safety documentation.

  • Drug-linker conjugate for ADC synthesis
  • High purity (approximately 98.5%) solid, white to light yellow
  • Molecular formula C56H73N9O15S and molecular weight 1,144.30
  • CAS number 2964513-44-6
  • Store at -80°C, protect from light, store under nitrogen

Catalog No. 50-002-04411 Supplier Medchemexpress LLC Supplier No. HY155870A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-01 formic is a drug-linker conjugate used in the synthesis of antibody-drug conjugates (ADCs) and related research applications. It is supplied as a high-purity solid for laboratory research use only and should be handled and stored according to the product datasheet and safety documentation.

  • Drug-linker conjugate for ADC synthesis
  • High purity (approximately 98.5%) solid, white to light yellow
  • Molecular formula C56H73N9O15S and molecular weight 1,144.30
  • CAS number 2964513-44-6
  • Store at -80°C, protect from light, store under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.