Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-01 formic | 2964513-44-6 | 98.5% | 1144.30 g/mol | C56H73N9O15S | 1 MG
SDP

Catalog No. 5000204331
Encompass_Preferred

DL-01 formic is a drug-linker conjugate intended for research use in the synthesis of antibody-drug conjugates (ADCs). Supplied as a white to light yellow solid, it has high purity and requires ultra-low temperature storage to preserve stability.

  • Drug-linker conjugate for antibody-drug conjugate (ADC) synthesis.
  • White to light yellow solid physical form.
  • Purity approximately 98.5%.
  • Molecular weight 1,144.30 g/mol.
  • CAS number 2964513-44-6.
  • Stored at -80°C and protected from light under inert gas.
  • Available in small milligram quantities for research use.

Catalog No. 50-002-04331 Supplier Medchemexpress LLC Supplier No. HY155870A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-01 formic is a drug-linker conjugate intended for research use in the synthesis of antibody-drug conjugates (ADCs). Supplied as a white to light yellow solid, it has high purity and requires ultra-low temperature storage to preserve stability.

  • Drug-linker conjugate for antibody-drug conjugate (ADC) synthesis.
  • White to light yellow solid physical form.
  • Purity approximately 98.5%.
  • Molecular weight 1,144.30 g/mol.
  • CAS number 2964513-44-6.
  • Stored at -80°C and protected from light under inert gas.
  • Available in small milligram quantities for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.