Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-1,2-Hexanediol, Spectrum™ Chemical
SDP

Catalog No. 18620856 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
1 kg
20 kg
30 kg
200 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-856 200 kg
18-620-852 100 g
18-620-853 1 kg
18-620-854 20 kg
18-620-855 30 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-620-856 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H2008200KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

Synonym: Hexane-1,2-Diol, 1,2-Dihydroxyhexane, 1,2-Hexanediol

DL-1,2-Hexanediol, also known as 1,6-Hexanediol or Hexamethylene glycol, can be used for polyurethane production and industrial polyester. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 6920-22-5
Molecular Weight (g/mol) 118.18

Specifications

Quantity 200 kg
Packaging Drum
Boiling Point 223°C to 224°C
Percent Purity 98%
Grade Ungraded
Chemical Name or Material DL-1,2-Hexanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.