Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-1-Amino-2-propanol, 94%, contains approx. 5% 2-Amino-1-propanol

Catalog No. p-57512
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC149650010 1 kg Glass bottle
AC149650025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC149650010 Supplier Thermo Scientific Chemicals Supplier No. 149650010
Only null left
Add to Cart

CAS: 78-96-6 | C3H9NO | 75.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-96-6
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00008139
InChI Key HXKKHQJGJAFBHI-UHFFFAOYNA-N
Synonym 1-amino-2-propanol, isopropanolamine, monoisopropanolamine, threamine, 2-propanol, 1-amino, 2-hydroxypropylamine, amino-2-propanol, mipa, 1-methyl-2-aminoethanol, 2-hydroxypropanamine
PubChem CID 4
ChEBI CHEBI:19030
IUPAC Name 1-aminopropan-2-ol
SMILES CC(O)CN

Specifications

Melting Point -2°C
Density 0.9600g/mL
Boiling Point 161°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 93% min. (GC)
Packaging Glass bottle
Refractive Index 1.4470 to 1.449
Linear Formula CH3CH(OH)CH2NH2
Quantity 1 kg
Beilstein 04, 289
Specific Gravity 0.96
Solubility Information Solubility in water: freely soluble
Viscosity 31 mPa.s (20°C)
Formula Weight 75.11
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material DL-1-Amino-2-propanol, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 201-162-7
RTECSNumber UA5775000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.