Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2,4-Pentanediol 97%, Thermo Scientific™

Catalog No. AC124390010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC124390010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC124390010 Supplier Thermo Scientific Chemicals Supplier No. 124390010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-69-4
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.149
InChI Key GTCCGKPBSJZVRZ-UHFFFAOYSA-N
Synonym 2,4-pentanediol, 2,4-amylene glycol, isoamylene alcohol, pentanediol-2,4, 2,4-amyleneglycol, dl-2,4-pentanediol, 2,4-pentanediol dl+meso, wln: qy&1yq, 2,4-pentandiol, 2,4-dihydroxypentane
PubChem CID 12262
IUPAC Name pentane-2,4-diol
SMILES CC(CC(C)O)O

Specifications

Density 0.95
Linear Formula CH3CH(OH)CH2CH(OH)CH3
Quantity 1 g
Beilstein 01, IV, 2543
Formula Weight 104.15
Chemical Name or Material DL-2, 4-Pentanediol, 97%
Hazard Category HYGROSCOPIC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.