Learn More
Cayman Chemical DL-2,6-dimethyl Heptanoic Acid, Purity: ≥98%, CAS: 7494-12-4, 25 mg
SDP
Catalog No. 503015691
Encompass_Preferred
An aliphatic branched acid; inhibits the growth of V. fischeri (EC50 = 17 µM); a building block in the synthesis of 2,6-dimethyl heptanoyl-CoA
Catalog No. 50-301-5691 Supplier Cayman Chemical Supplier No. 2482225 MG
An aliphatic branched acid; inhibits the growth of V. fischeri (EC50 = 17 µM); a building block in the synthesis of 2,6-dimethyl heptanoyl-CoA