Learn More
Cayman Chemical DL-2 6-dimethyl Heptanoic Aci
SDP
Supplier:  Cayman Chemical 2482250 MG
Encompass_Preferred
An aliphatic branched acid; inhibits the growth of V. fischeri (EC50 = 17 µM); a building block in the synthesis of 2,6-dimethyl heptanoyl-CoA
Catalog No. 50-301-5872