Learn More
Cayman Chemical DL-2 6-dimethyl Heptanoic Aci
SDP
Catalog No. 503015862
Encompass_Preferred
An aliphatic branched acid; inhibits the growth of V. fischeri (EC50 = 17 µM); a building block in the synthesis of 2,6-dimethyl heptanoyl-CoA
Catalog No. 50-301-5862 Supplier Cayman Chemical Supplier No. 2482210 MG
An aliphatic branched acid; inhibits the growth of V. fischeri (EC50 = 17 µM); a building block in the synthesis of 2,6-dimethyl heptanoyl-CoA