Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Amino-1-propanol 98.0+%, TCI America™
SDP

Catalog No. A09425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09425G 5 g
A094225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09425G Supplier TCI America Supplier No. A09425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6168-72-5
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00008085
InChI Key BKMMTJMQCTUHRP-UHFFFAOYNA-N
Synonym dl-alaninol, 2-aminopropanol, 2-amino-1-propanol, dl-2-amino-1-propanol, 1-propanol, 2-amino, 1-hydroxy-2-aminopropane, 2-amino-2-methylethanol, alaninol, propanol, 2-amino, 1-methyl-2-hydroxyethylamine
PubChem CID 5126
IUPAC Name 2-aminopropan-1-ol
SMILES CC(N)CO

Specifications

Color Yellow
Boiling Point 175°C
Quantity 5 g
UN Number 2735
Formula Weight 75.11
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material DL-2-Amino-1-propanol
EINECSNumber (2)-0323&(9)-0122
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.