Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-2-amino-3-phosphonopropionic acid | 5652-28-8 | MFCD00014350 | 98.0% | 169.07 g/mol | C3H8NO5P | 10 MG
SDP

Catalog No. 5000212172
Encompass_Preferred

DL-AP3 (DL-2-amino-3-phosphonopropionic acid) is a small-molecule research compound used to probe glutamatergic signaling. It functions as a competitive antagonist of group I metabotropic glutamate receptors and inhibits phosphoserine phosphatase, with reported neuroprotective effects in cellular studies.

  • Competitive antagonist of mGluR1 and mGluR5.
  • Inhibits phosphoserine phosphatase.
  • Reported neuroprotective effects in cellular models.
  • High purity suitable for research (98.0%).
  • Water solubility: 5 mg/mL with ultrasonic warming to 60°C.
  • Storage: -20 °C (solid); in solution -80 °C up to 6 months.
  • Molecular weight 169.07 g/mol; formula C3H8NO5P; CAS 5652-28-8.

Catalog No. 50-002-12172 Supplier Medchemexpress LLC Supplier No. HY10078610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-AP3 (DL-2-amino-3-phosphonopropionic acid) is a small-molecule research compound used to probe glutamatergic signaling. It functions as a competitive antagonist of group I metabotropic glutamate receptors and inhibits phosphoserine phosphatase, with reported neuroprotective effects in cellular studies.

  • Competitive antagonist of mGluR1 and mGluR5.
  • Inhibits phosphoserine phosphatase.
  • Reported neuroprotective effects in cellular models.
  • High purity suitable for research (98.0%).
  • Water solubility: 5 mg/mL with ultrasonic warming to 60°C.
  • Storage: -20 °C (solid); in solution -80 °C up to 6 months.
  • Molecular weight 169.07 g/mol; formula C3H8NO5P; CAS 5652-28-8.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.