Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-2-aminooctanoic acid | 644-90-6 | MFCD00008102 | 98.0% | 159.23 g/mol | C8H17NO2 | 500 MG
SDP

Catalog No. 5000303293
Encompass_Preferred

2-Aminooctanoic acid is a small, aliphatic amino acid (DL mixture) used as a research reagent for peptide modification and biochemical studies. It features a primary amino group at the 2-position and is applicable for synthesis, analytical work, and modification of antimicrobial peptides to probe activity and stability.

  • High purity (98.0%) suitable for research applications.
  • Molecular formula C8H17NO2 and molecular weight 159.23 g/mol.
  • CAS number 644-90-6 for unambiguous identification.
  • Can be coupled at the N- or C-terminus for peptide modification.
  • Supplied in laboratory-scale pack sizes for synthetic and analytical use.

Catalog No. 50-003-03293 Supplier Medchemexpress LLC Supplier No. HYW015461500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-Aminooctanoic acid is a small, aliphatic amino acid (DL mixture) used as a research reagent for peptide modification and biochemical studies. It features a primary amino group at the 2-position and is applicable for synthesis, analytical work, and modification of antimicrobial peptides to probe activity and stability.

  • High purity (98.0%) suitable for research applications.
  • Molecular formula C8H17NO2 and molecular weight 159.23 g/mol.
  • CAS number 644-90-6 for unambiguous identification.
  • Can be coupled at the N- or C-terminus for peptide modification.
  • Supplied in laboratory-scale pack sizes for synthetic and analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.