Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Hydroxybutyric Acid (contains Polymolecular esterification product) 95.0+%, TCI America™
SDP

Catalog No. H022725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H022725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H022725G Supplier TCI America Supplier No. H022725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 600-15-7
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD01318568,MFCD00070502
InChI Key AFENDNXGAFYKQO-UHFFFAOYNA-N
PubChem CID 11266
ChEBI CHEBI:1148
IUPAC Name 2-hydroxybutanoic acid
SMILES CCC(O)C(O)=O

Specifications

Melting Point 43°C
Color White
Boiling Point 260°C
Quantity 25 g
Formula Weight 104.11
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-2-Hydroxybutyric Acid (contains Polymolecular esterification product)
EINECSNumber (2)-1429
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.