Learn More
Medchemexpress LLC DL-2-methylbutyryl-L-carnitine-d3 (chloride) | 00-00-0 | MFCD34676379 | 99.2% | C12H21D3ClNO4 | 10MG
SDP
Catalog No. 5000258255
Encompass_Preferred
2-Methylbutyryl-L-carnitine-d3 (chloride) is a deuterium-labeled isotopic standard of 2-methylbutyryl-L-carnitine supplied as the chloride salt. It is intended for use as an internal standard or tracer in quantitative mass spectrometry and NMR workflows, supporting reliable peak identification and quantification.
  • Deuterium-labeled (d3) compound with three deuterium atoms.
  • High chemical purity (≈99.2%) as reported on manufacturer COA.
  • Provided as the chloride salt for improved solubility.
  • Suitable as internal standard for LC-MS, GC-MS, and NMR quantitation.
  • Recommended storage at -80 °C for long-term stability.
Catalog No. 50-002-58255 Supplier Medchemexpress LLC Supplier No. HY132958S10MG
2-Methylbutyryl-L-carnitine-d3 (chloride) is a deuterium-labeled isotopic standard of 2-methylbutyryl-L-carnitine supplied as the chloride salt. It is intended for use as an internal standard or tracer in quantitative mass spectrometry and NMR workflows, supporting reliable peak identification and quantification.
  • Deuterium-labeled (d3) compound with three deuterium atoms.
  • High chemical purity (≈99.2%) as reported on manufacturer COA.
  • Provided as the chloride salt for improved solubility.
  • Suitable as internal standard for LC-MS, GC-MS, and NMR quantitation.
  • Recommended storage at -80 °C for long-term stability.