Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-3,4-Dihydroxymandelic Acid 98.0+%, TCI America™
SDP

Catalog No. D0582100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0582100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D0582100MG Supplier TCI America Supplier No. D0582100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 775-01-9
Molecular Formula C8H8O5
Molecular Weight (g/mol) 184.147
MDL Number MFCD00004231
InChI Key RGHMISIYKIHAJW-UHFFFAOYSA-N
PubChem CID 85782
ChEBI CHEBI:27637
IUPAC Name 2-(3,4-dihydroxyphenyl)-2-hydroxyacetic acid
SMILES C1=CC(=C(C=C1C(C(=O)O)O)O)O

Specifications

Melting Point 131°C
Quantity 100 mg
Formula Weight 184.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-3,4-Dihydroxymandelic Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.