Learn More
Cayman Chemical DL-3 4-Dihydroxyphenyl glycol
SDP
Catalog No. 502952088
Encompass_Preferred
An active metabolite of norepinephrine; scavenges DPPH radicals in a cell-free assay (IC50 = 1.55 µM); reduces LPS-induced nitrite production in isolated mouse peritoneal macrophages at 50 and 100 µM; decreases increases in iNOS and COX-2 levels induced by LPS in isolated mouse macrophages at 50 and 100 µM; inhibits LPS-induced degradation of IκBα in isolated mouse macrophages
Catalog No. 50-295-2088 Supplier Cayman Chemical Supplier No. 117081 MG
An active metabolite of norepinephrine; scavenges DPPH radicals in a cell-free assay (IC50 = 1.55 µM); reduces LPS-induced nitrite production in isolated mouse peritoneal macrophages at 50 and 100 µM; decreases increases in iNOS and COX-2 levels induced by LPS in isolated mouse macrophages at 50 and 100 µM; inhibits LPS-induced degradation of IκBα in isolated mouse macrophages