Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-3-Aminobutyric Acid 98.0+%, TCI America™
SDP

Catalog No. A02815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A02815G 5 g
A02811G 1 g
A028125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A02815G Supplier TCI America Supplier No. A02815G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 541-48-0 | C4H9NO2 | 103.121 g/mol

Chemical Identifiers

CAS 541-48-0
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00008087
InChI Key OQEBBZSWEGYTPG-UHFFFAOYSA-N
Synonym dl-3-aminobutyric acid, 3-aminobutyric acid, beta-aminobutyric acid, butanoic acid, 3-amino, dl-3-amino-n-butyric acid, baba, 3-amino-butyric acid, 3-methyl-beta-alanine, butyric acid, 3-amino, dl-.beta.-aminobutyric acid
PubChem CID 10932
ChEBI CHEBI:37081
IUPAC Name 3-aminobutanoic acid
SMILES CC(CC(=O)O)N

Specifications

Melting Point 194°C
Color White
Quantity 5 g
Formula Weight 103.12
Percent Purity ≥98.0% (T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material DL-3-Aminobutyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.