Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich DL-3-aminoisobutyric acid monohydrate

Catalog No. CDS014136 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS014136 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS014136 Supplier Sigma Aldrich Supplier No. CDS014136250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,n-dimethyltetradecylamine has been used for the synthesis of 1,6-bis(n,n-tetradecyl dimethylammonium adipate), a counterion coupled gemini (cocogem) surfactant. it may also be used in the following studies:
  • For the isolation of recombinant botulinum neurotoxin fragments in the yeast pichia pastoris by cell permeabilization method
  • Synthesis of gemini surfactants belonging to the series of alkanediyl-α,-d-bis-(dimethylalkyl ammonium bromide, which were evaluated as corrosion inhibitors of iron in hydrochloric acid medium
  • Preparation of cationic gemini surfactant of the type n-alkanediyl 1,2-ethane bis(dimethyl ammonium bromide)

Specifications

Molecular Formula C4H11NO3
Linear Formula C4H11NO3
MDL Number MFCD00149561
Quantity 100 mL
Molecular Weight (g/mol) 121.14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.