Learn More
AdipoGen DL-3-Hydroxybutyric acid Na
SDP
Catalog No. 502560761
Encompass_Preferred
Chemical. CAS 150-83-4. Formula C4H7NaO3. MW 126.1. Synthetic. Neuroprotective compound in experimental models of neurodegenerative diseases. Shown to be oxidized as an energetic substrate in the brain during starvation. May inhibit brain glucose oxidation, which may account for the suppression of hepatic glucose production observed after administration of ketone bodies in humans.
Catalog No. 50-256-0761 Supplier AdipoGen Supplier No. CDXH0080G001
Chemical. CAS 150-83-4. Formula C4H7NaO3. MW 126.1. Synthetic. Neuroprotective compound in experimental models of neurodegenerative diseases. Shown to be oxidized as an energetic substrate in the brain during starvation. May inhibit brain glucose oxidation, which may account for the suppression of hepatic glucose production observed after administration of ketone bodies in humans.